(±)-Methyl-2-bromopropionate ≥97%

Supplier: BeanTown Chemical

Synonyms: Methyl 2-bromopropanoate

128390-500G 128390-25G 128390-100G
BT128390-500GEA 177.6 USD
BT128390-500G BT128390-25G BT128390-100G
(±)-Methyl-2-bromopropionate ≥97%
(±)-Methyl 2-bromopropionate

  • Refractive index: 1.451
  • Liquid

Formula: C₄H₇BrO₂
MW: 167 g/mol
Boiling Pt: 51 °C/19 mmHg
Density: 1.497 g/mL
Flash Pt: 51 °C (123 °F)
MDL Number: MFCD00000143
CAS Number: 5445-17-0
EINECS: 226-642-3
ADR: 3(8),III

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR