Dicyclohexylamine nitrite ≥97%

Supplier: BeanTown Chemical

Synonyms: N,N-Dicyclohexylammonium nitrite

128170-100G
BT128170-100GEA 75.31 USD
BT128170-100G
Dicyclohexylamine nitrite ≥97%
Dicyclohexylamine nitrite

  • RTECS: HY4200000
  • Powder

Formula: C₁₂H₂₃N
MW: 228.34 g/mol
Melting Pt: 182…183 °C (dec.)
MDL Number: MFCD00013128
CAS Number: 3129-91-7
EINECS: 221-515-9
ADR: 4.1,III

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR