Ammonium hexafluorosilicate ≥98%
Supplier: BeanTown Chemical
128075-5KG
128075-250G
128075-1KG
BT128075-1KGEA
85.18
USD
BT128075-1KG
BT128075-250G
BT128075-5KG
Ammonium hexafluorosilicate ≥98%
Ammonium hexafluorosilicate
- Powder
Formula:
H₈F₆N₂Si MW: 178.15 g/mol Density: 2.01 g/mL |
MDL Number:
MFCD00010887 CAS Number: 16919-19-0 EINECS: 240-968-3 UN: 2854 ADR: 6.1,III |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...