1-(2,4-Difluorophenyl)piperazine ≥99%

Supplier: BeanTown Chemical

128040-25G 128040-5G 128040-1G
BT128040-1GEA 37.6 USD
BT128040-1G BT128040-5G BT128040-25G
1-(2,4-Difluorophenyl)piperazine ≥99%
1-(2,4-Difluorophenyl)piperazine

  • Air Sensitive
  • Crystalline/Powder

Formula: C₁₀H₁₂F₂N₂
MW: 198.22 g/mol
MDL Number: MFCD00082588
CAS Number: 115761-79-0
UN: 2923
ADR: 8,II

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR