Dimagnesium silicide ≥99.99% (trace metals basis)

Supplier: BeanTown Chemical

Synonyms: Magnesium silicide

126920-50G 126920-250G 126920-10G
BT126920-50GEA 244.95 USD
BT126920-50G BT126920-250G BT126920-10G
Dimagnesium silicide ≥99.99% (trace metals basis)
Dimagnesium silicide

  • Air Sensitive, Moisture Sensitive
  • 3-12mm Pieces

Formula: Mg₂Si
MW: 76.70 g/mol
Melting Pt: 1102 °C
Density: 2 g/mL
EINECS: 245-254-5
UN: 2624
ADR: 4.3,II

Order Now

Specification Test Results

Assay 99.99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR