2-Chloroethyl chloroformate ≥97%

Supplier: BeanTown Chemical

Synonyms: 2-Chloroethyl carbonochloridate

126865-100G
BT126865-100GEA 128.02 USD
BT126865-100G
2-Chloroethyl chloroformate ≥97%
2-Chloroethyl chloroformate

  • RTECS: LQ5950000
  • Refractive index: 1.446
  • Moisture Sensitive
  • Liquid

Formula: C₃H₄Cl₂O₂
MW: 142.9687 g/mol
Boiling Pt: 155…156 °C
Density: 1.385 g/mL
Flash Pt: 70 °C (158 °F)
EINECS: 210-982-4
ADR: 6.1 (8),II

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR