2-Mercaptoethanol ≥98%

Synonyms: 2-Hydroxyethyl mercaptan1,1,2,2-tetradeuterio-1-deuteriooxy-2-deuteriosulfanylethanebeta-Mercaptoethanolβ-Mercaptoethanolβ-Hydroxyethanethiol

126185-1KG 126185-250G 126185-5KG
BT126185-250GEA 27.83 USD
BT126185-250G BT126185-1KG BT126185-5KG
2-Mercaptoethanol ≥98%
2-Mercaptoethanol

  • RTECS: KL5600000
  • Refractive index: 1.500
  • Air Sensitive, Hygroscopic
  • Liquid

Formula: HSCH₂CH₂OH
MW: 78.14 g/mol
Boiling Pt: 157 °C
Density: 1.114 g/mL
Flash Pt: 73 °C (163 °F)
MDL Number: MFCD00004890
CAS Number: 60-24-2
EINECS: 200-464-6
UN: 2966
ADR: 6.1,II
https://us.vwr.com/stibo/web/std.lang.all/79/24/2297924.jpg

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR