3-Bromopyridine ≥98%

Supplier: BeanTown Chemical

126140-100G 126140-500G 126140-25G
BT126140-25GEA 47.67 USD
BT126140-25G BT126140-500G BT126140-100G
3-Bromopyridine ≥98%
3-Bromopyridine

  • Refractive index: 1.571
  • Liquid

Formula: C₅H₄BrN
MW: 158 g/mol
Boiling Pt: 173 °C
Density: 1.64 g/mL
Flash Pt: 52 °C (126 °F)
MDL Number: MFCD00006373
CAS Number: 626-55-1
EINECS: 210-952-0
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR