Tripropyltin chloride ≥95%
Supplier: BeanTown Chemical
Synonyms:
Tri-n-propyltin chloride
, Chlorotripropylstannane
126065-25G
126065-5G
126065-1G
BT126065-25GEA
609.72
USD
BT126065-25G
BT126065-1G
BT126065-5G
Tripropyltin chloride ≥95%
Tripropyltin chloride
- Refractive index: 1.491
- Moisture Sensitive
- Liquid
Formula:
C₉H₂₁ClSn MW: 283.43 g/mol Boiling Pt: 123 °C/13 mmHg Melting Pt: –23.5 °C Density: 1.268 g/mL |
MDL Number:
MFCD00013610 CAS Number: 2279-76-7 EINECS: 218-910-3 UN: 2788 ADR: 6.1,II |
Specification Test Results
Assay | 95% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...