Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

o-Xylene ≥96.0% for spectrophotometry

Supplier: BeanTown Chemical

Synonyms: 1,2-Dimethylbenzene

125645-4X1L 125645-1L 125645-4L
BT125645-4X1LEA 243.18 USD InStock
BT125645-4X1L BT125645-1L BT125645-4L
o-Xylene ≥96.0% for spectrophotometry
o-Xylene
CAS: 95-47-6; EC No: 202-422-2; MDL No: MFCD00008519; RTECS: ZE2450000
UN No: UN1307; Haz Class: 3; Packing Group: II
Liquid; Linear Formula: C6H4(CH3)2; Molecular Formula: C8H10O; MW: 106.17
Melting Point: -26 - -23°; Boiling Point: 143-145°; Flash point: 31°C (89°F)
Density (g/mL): 0.879; Refractive Index: 1.505


Formula: C₈H₁₀
MW: 106,17 g/mol
Boiling Pt: 144 °C (1013 hPa)
Melting Pt: -25 °C
Density: 0.88 g/cm³ (20 °C)
Flash Pt: 17 °C (closed cup)
Storage Temperature: ambient
MDL Number: MFCD00008519
CAS Number: 95-47-6
EINECS: 202-422-2
UN: 1307
ADR: 3,III
Merck Index: 12,10214
https://us.vwr.com/stibo/web/std.lang.all/85/44/768544.jpg
Order Now
Specification Test Results
Assay 96.0% min.


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty