4-(Chloromethyl)phenyltrimethoxysilane ≥90%, Technical Grade
Supplier: BeanTown Chemical
125635-5G
125635-100G
125635-25G
BT125635-5GEA
39.58
USD
BT125635-5G
BT125635-25G
BT125635-100G
4-(Chloromethyl)phenyltrimethoxysilane ≥90%, Technical Grade
4-(Chloromethyl)phenyltrimethoxysilane
- Moisture Sensitive
- Liquid
Formula:
C₁₀H₁₅ClO₃Si Boiling Pt: 134…143 °C/10 mmHg Density: 1.12 g/mL Flash Pt: 81 °C (177 °F) |
MDL Number:
MFCD00045269 CAS Number: 24413-04-5 EINECS: 246-235-4 ADR: 8,II |
Specification Test Results
Assay | 90% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...