Isobutyl methacrylate ≥99.5% stabilized

Synonyms: Methacrylic acid Isobutyl ester

125445-4L 125445-250ML 125445-1L
BT125445-1LEA 59.16 USD
BT125445-1L BT125445-4L BT125445-250ML
Isobutyl methacrylate ≥99.5% stabilized
Isobutyl methacrylate

  • RTECS: OZ4900000
  • Liquid

Formula: C₈H₁₄O₂
MW: 142.2 g/mol
Boiling Pt: 155 °C
Density: 0.889 g/mL
Flash Pt: 46 °C (115 °F)
MDL Number: MFCD00008931
CAS Number: 97-86-9
EINECS: 202-613-0
UN: 2283
ADR: 3,III

Order Now

Specification Test Results

Assay 99.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR