5-Cyano-1-pentyne ≥98%
Supplier: BeanTown Chemical
Synonyms:
5-Hexynenitrile
125225-5G
125225-1G
BT125225-5GEA
97.12
USD
BT125225-5G
BT125225-1G
5-Cyano-1-pentyne ≥98%
5-Cyano-1-pentyne
- Refractive index: 1.44
- Liquid
Formula:
C₆H₇N MW: 93.13 g/mol Boiling Pt: 115…117 °C Density: 0.889 g/mL Flash Pt: 42 °C (108 °F) |
MDL Number:
MFCD00001978 CAS Number: 14918-21-9 UN: 1992 ADR: 3,III |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...