1,1,1,3,3,3-Hexafluoro-2-phenyl-2-propanol ≥98%

Supplier: BeanTown Chemical

Synonyms: 1,1,1,3,3,3-Hexafluoro-2-phenylpropan-2-ol2,2,2,2',2',2'-Hexafluorocumyl Alcoholalpha,alpha-Bis(trifluoromethyl)benzyl Alcohol

124650-5G 124650-1G 124650-25G
BT124650-5GEA 60.98 USD
BT124650-5G BT124650-25G BT124650-1G
1,1,1,3,3,3-Hexafluoro-2-phenyl-2-propanol ≥98%
1,1,1,3,3,3-Hexafluoro-2-phenyl-2-propanol

  • Refractive index: 1.415
  • Liquid

Formula: C₉H₆F₆O
MW: 244.14 g/mol
Boiling Pt: 160 °C
Density: 1.45 g/mL
Flash Pt: 60 °C (140 °F)
MDL Number: MFCD00040983
CAS Number: 718-64-9
EINECS: 211-943-4
ADR: 3,III

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR