1,4-Diethynylbenzene ≥97%

Supplier: BeanTown Chemical

Synonyms: p-Diethynylbenzene1,4-Diethylnylbenzene

124490-5G
BT124490-5GEA 174.49 USD
BT124490-5G
1,4-Diethynylbenzene ≥97%
1,4-Diethynylbenzene

  • RTECS: CZ5643000
  • Crystalline/Powder

Formula: C₁₀H₆
MW: 126.16 g/mol
Melting Pt: 94…98 °C
MDL Number: MFCD00078375
CAS Number: 935-14-8
ADR: 4.1,III

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR