N-Methylisobutylamine ≥98%

Supplier: BeanTown Chemical

124140-5G 124140-1G
BT124140-1GEA 18.74 USD
BT124140-1G BT124140-5G
N-Methylisobutylamine ≥98%
N-Methylisobutylamine

  • Liquid

Formula: C₅H₁₃N
Boiling Pt: 78 °C
Density: 0.73 g/mL
MDL Number: MFCD00015043
CAS Number: 625-43-4
EINECS: 210-893-0
ADR: 3 (8),II

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR