5-Bromo-8-nitroisoquinoline ≥90%

Supplier: BeanTown Chemical

123550-1G
BT123550-1GEA 64.81 USD
BT123550-1G
5-Bromo-8-nitroisoquinoline ≥90%
5-Bromo-8-nitroisoquinoline

  • Solid

Formula: C₉H₅BrN₂O₂
MW: 253.0500 g/mol
Melting Pt: 136…140 °C
MDL Number: MFCD02091227
CAS Number: 63927-23-1
ADR: 6.1,III

Order Now

Specification Test Results

Assay 90%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR