Pentafluoropropionic acid ≥97%
Supplier: BeanTown Chemical
Synonyms:
Perfluoropropionic acid
, Perfluoropropanoic acid
, 2,2,3,3,3-Pentafluoropropionic acid
, Pentafluoropropanoic acid
123490-25G
123490-100G
123490-500G
BT123490-25GEA
69.8
USD
BT123490-25G
BT123490-100G
BT123490-500G
Pentafluoropropionic acid ≥97%
Pentafluoropropionic acid
- RTECS: UF6475000
- Refractive index: 1.284
- Hygroscopic
- Liquid
Formula:
C₃HF₅O₂ MW: 164.03 g/mol Boiling Pt: 95…97 °C Density: 1.561 g/mL |
MDL Number:
MFCD00004170 CAS Number: 422-64-0 EINECS: 207-021-6 UN: 3265 ADR: 8,II |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...