Phenyl isocyanate ≥98%

123385-250G 123385-100G
BT123385-250GEA 70.72 USD
BT123385-250G BT123385-100G
Phenyl isocyanate ≥98%
Phenyl isocyanate

  • RTECS: DA3675000
  • Refractive index: 1.535
  • Moisture Sensitive
  • Liquid

Formula: C₇H₅NO
MW: 119.12 g/mol
Boiling Pt: 162…163 °C
Melting Pt: –30 °C
Density: 1.096 g/mL
Flash Pt: 51 °C (123 °F)
MDL Number: MFCD00001994
CAS Number: 103-71-9
EINECS: 203-137-6
UN: 2487
ADR: 6.1,I

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR