5-Bromo-2-isopropoxybenzonitrile ≥97%

Supplier: BeanTown Chemical

123365-250MG 123365-1G
BT123365-250MGEA 62.42 USD
BT123365-250MG BT123365-1G
5-Bromo-2-isopropoxybenzonitrile ≥97%
5-Bromo-2-isopropoxybenzonitrile

  • Powder

Formula: C₁₀H₁₀BrNO
MW: 240.10 g/mol
MDL Number: MFCD02257441
CAS Number: 515832-52-7
ADR: 6.1,III

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR