Lithium fluoride ≥99.5%
Supplier: BeanTown Chemical
123100-100G
123100-5G
123100-25G
BT123100-100GEA
675.15
USD
BT123100-100G
BT123100-5G
BT123100-25G
Lithium fluoride ≥99.5%
Lithium fluoride
- Broken Crystals
Formula:
FLi MW: 25.94 g/mol Boiling Pt: 1673 °C Melting Pt: 845 °C Density: 2.64 g/mL |
MDL Number:
MFCD00011090 CAS Number: 7789-24-4 EINECS: 232-152-0 UN: 3288 ADR: 6.1,III |
Specification Test Results
Assay | 99.5% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...