1-Chlorooctane ≥99%

Supplier: BeanTown Chemical

Synonyms: n-Octyl chloride

122725-100ML 122725-500ML
BT122725-100MLEA 22.55 USD
BT122725-100ML BT122725-500ML
1-Chlorooctane ≥99%
1-Chlorooctane

  • Refractive index: 1.430
  • Liquid

Formula: C₈H₁₇Cl
MW: 148.68 g/mol
Boiling Pt: 183 °C
Melting Pt: –61 °C
Density: 0.875 g/mL
Flash Pt: 61 °C (142 °F)
MDL Number: MFCD00001022
CAS Number: 111-85-3
EINECS: 203-915-5
UN: 3082
ADR: 9,III

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR