BEAN122470

122470-2.5L
BT122470-2.5LEA 64.97 USD
BT122470-2.5L
BEAN122470
  • RTECS: NQ4025000
  • Refractive index: 1.372
  • Air Sensitive
  • Liquid
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR