Heptanenitrile ≥98%

Supplier: BeanTown Chemical

Synonyms: Hexyl Cyanide

122380-5G 122380-25G
BT122380-25GEA 135.01 USD
BT122380-25G BT122380-5G
Heptanenitrile ≥98%
Heptanenitrile

  • Liquid

Formula: C₇H₁₃N
MW: 111.19 g/mol
Boiling Pt: 186…187 °C
Density: 0.81 g/mL
Flash Pt: 58 °C (136 °F)
MDL Number: MFCD00039503
CAS Number: 629-08-3
EINECS: 211-071-4
ADR: 3 (6.1),III

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR