Tris(trimethylsilyl)silane ≥97%

Supplier: BeanTown Chemical

Synonyms: 1,1,1,3,3,3-Hexamethyl-2-(trimethylsilyl)trisilaneTTMSS

122050-25G 122050-1G 122050-5G
BT122050-25GEA 257.57 USD
BT122050-25G BT122050-1G BT122050-5G
Tris(trimethylsilyl)silane ≥97%
Tris(trimethylsilyl)silane

  • Refractive index: 1.489
  • Light Sensitive
  • Liquid

Formula: C₉H₂₈Si₄
MW: 248.66 g/mol
Boiling Pt: 73 °C/5 mmHg
Density: 0.806 g/mL
Flash Pt: 55 °C (131 °F)
MDL Number: MFCD00077893
CAS Number: 1873-77-4
ADR: 3,II

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR