Diphenylamine ≥98%

Supplier: BeanTown Chemical

Synonyms: N-PhenylbenzeneamineN-Phenylaniline

122015-100G 122015-2.5KG 122015-500G
BT122015-500GEA 44.68 USD
BT122015-500G BT122015-2.5KG BT122015-100G
Diphenylamine ≥98%
Diphenylamine

  • RTECS: JJ7800000
  • Air Sensitive, Light Sensitive
  • Solid

Formula: (C₆H₅)₂NH
MW: 169.23 g/mol
Boiling Pt: 302 °C
Melting Pt: 50…53 °C
Flash Pt: 153 °C (307 °F)
MDL Number: MFCD00003014
CAS Number: 122-39-4
EINECS: 204-539-4
UN: 3077
ADR: 9,III
https://us.vwr.com/stibo/web/std.lang.all/76/36/2297636.jpg

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR