Ready Reactor, Kilo Scale, Ace Glass Incorporated
10001-201 10001-501
10767-952EA 29855.09 USD
10767-952 10767-954
Ready Reactor, Kilo Scale, Ace Glass Incorporated
Pre-packaged, turnkey 20 and 50 L reaction systems for research and production purposes.
Systems feature jacketed vessels, multi-neck flask heads, universal support stand, and a rod-mounted brushless DC stir motor in the 20 L (IKA Eurostar 60 Control), or a flange-mounted 3/8 hp brushless DC motor and 120V control in the 50 L.