Paraffin, PELORIS Parablocks™
Supplier: Leica Biosystems
Synonyms: Wax ParaffinParaffin wax
39V1001
10015-048CS 224.2 USD
10015-048
Paraffin, PELORIS Parablocks™
Paraffin
Leica PELORIS Parablocks™ are pre-formed blocks of paraffin that are provided in plastic molds.
  • Provided in plastic molds
  • Reduced risk of pellet spills
  • Eliminates need to transfer molten paraffin
The preformed blocks fit easily into the paraffin chamber of the Leica PELORIS Tissue Processor and will melt faster than pellets or flakes. Once the chamber is filled with the solid paraffin blocks, there is no need for the technician to return later to determine the paraffin volume and add additional paraffin. The format reduces risks as the solid blocks eliminate pellet spills and the need to transfer molten paraffin.
CAS Number: 8002-74-2