Frostbite® Rapid Coolant, Leica
Supplier: Leica
3803100
10015-100CS 135.41 USD
10015-100
Frostbite® Rapid Coolant, Leica
Cooling Packs
New ozone safe Frostbite® Rapid Coolant does not contain CFCs, but still retains the ability to rapidly cool paraffin blocks and tissue.
  • Nonflammable
  • Reduced excess spray coverage
  • Ozone-safe
  • Rapidly cools paraffin blocks and tissue
Two quick bursts are all that is needed to ensure deeper penetration into both paraffin blocks and fresh tissue specimens. The trigger nozzle and needle tube help to localize the spray, thus reducing excess spray coverage and eliminating waste. Frostbite® Rapid Coolant is nonflammable.