Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PrimaCell™, Rat Renal Tubular Epithelial Cell Culture Kit, CHI Scientific

Supplier: CHI Scientific
2-82562
The PrimaCell™ system has been developed for the acquisition and growth of primary cells from a variety of different tissue types. Each PrimaCell™ kit has been optimized for each cell type to produce 4-7 times the number of primary cells obtained from published literature protocols. Each kit comes with 100 ml of tissue washing medium, our optimized tissue dissociation system OptiTDS™, 500 ml of growth medium, and enough growth supplements and serum to add to the supplied medium. Kits that require a Fibroblast control system also come with our FibrOut™ kit (added to the growth medium) which reduces or eliminates Fibroblast growth after 2-3 cell growth cycles.

The PrimaCell™ system has been developed for the acquisition and growth of primary cells from a variety of different tissue types

Stability/Storage: The reagents contained in this system are packed in dry ice or gel ice packs for shipping but the kit should be used immediately or the package should be stored at -20°C upon arrival. The reagents contained in this kit are stable and effective for up to 6 months at -20°C.

This kit includes (and is composed of) all of the items listed under the Accessories Section. Individual accessory items may be purchased instead of the full kit or in addition to the full kit as desired.

Each PrimaCell™ kit has been optimized for each cell type to produce 4-7 times the number of primary cells obtained from published literature protocols. Each kit comes with 100 ml of tissue washing medium, our optimized tissue dissociation system OptiTDS™, 500 ml of growth medium, and enough growth supplements and serum to add to the supplied medium. Kits that require a Fibroblast control system also come with our FibrOut™ kit (added to the growth medium) which reduces or eliminates Fibroblast growth after 2-3 cell growth cycles.



 

ORDER

Expand All / Collapse All
Description Size Supplier No. VWR Catalog Number Unit Price Quantity
Rat Kidney PrimaCell™ 5: Normal Renal Tubular Epithelial Cells Kit 2-82562 10784-288 Each Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

ACCESSORIES

Expand All / Collapse All

Accessories
Description Size Supplier No. VWR Catalog Number Unit Price Quantity
Rat Kidney FibrOut™ 5: Fibroblast Inhibitory System (for 500 ml medium) Kit 7-15242 10784-292 Each Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Rat Kidney OptiTDS™ 5: Tissue Dissociation System Kit 4-28269 10784-290 Each Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Rat Kidney PrimaCell™ 5: Normal Renal Tubular Epithelial Cells Growth Medium 5 x 100 mL 9-25062 10784-294 Each Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Rat Kidney PrimaCell™ 5: Normal Renal Tubular Epithelial Cells Growth Supplements with Serum (for 500 ml medium) 50 mL + 5 x 1 mL 9-26062 10784-296 Each Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Rat Kidney Tissue Preparation Buffer 5: Normal Renal Tubular Epithelial Cells 100 mL 9-80269 10784-298 Each Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping