Calibra® 832 Digital Macro Pipettes, DWK Life Sciences
Supplier: DWK Life Sciences
851340 851345
80078-624CS 957.16 USD
80078-624 53500-001
Calibra® 832 Digital Macro Pipettes, DWK Life Sciences
Pipettes Micropipettes
The Calibra® 832 digital macro pipette combines instant volume adjustment, mechanical precision, and ease of use to create an extremely reliable and robust macro pipette line.
This CE certified IVD 98/79 EEC pipette features winding-free, instant volume adjustment, shock/UV light, and autoclave resistance. All features combine to ensure long term performance and calibration stability.
Certifications: CE certified.
Ordering information: Pipette is supplied with pipette tip samples, individual QC certificate, and silicone lubricant. Pipette is also supplied with a three-year warranty.