Ziprasidone hydrochloride monohydrate ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 5-[2-[4-(1,2-Benzisothiazol-3-yl)-1-piperazinyl]ethyl]-6-chloro-1,3-dihydro-2H-indol-2-one Hydrochloride

Z0032-250MG Z0032-50MG
TCZ0032-250MGEA 165.45 USD
TCZ0032-250MG TCZ0032-50MG
Ziprasidone hydrochloride monohydrate ≥98.0% (by HPLC)
Ziprasidone hydrochloride monohydrate

CAS Number: 138982-67-9
MDL Number: MFCD00921885
Molecular Formula: C21H21ClN4OS
Molecular Weight: 449.39
Purity/Analysis Method: 98.0% (HPLC)
Form: Crystal
Color: White


Product Categories: Bioscience, Neurotransmitter Agent Ingredients
Other Product Names / Synonyms: 5-[2-[4-(1,2-Benzisothiazol-3-yl)-1-piperazinyl]ethyl]-6-chloro-1,3-dihydro-2H-indol-2-one Hydrochloride

Formula: C₂₁H₂₁ClN₄OS.HCl.H₂O
MW: 449.39 g/mol (anhy)
MDL Number: MFCD00921885
CAS Number: 138982-67-9
https://us.vwr.com/stibo/web/std.lang.all/74/50/28997450.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR