Xylobiose ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 4-O-beta-D-Xylopyranosyl-D-xylose

X0067-1G X0067-100MG
TCX0067-100MGEA 315.89 USD
TCX0067-100MG TCX0067-1G
Xylobiose ≥98.0% (by HPLC)
Xylobiose

CAS Number: 6860-47-5
MDL Number: MFCD00135984
Molecular Formula: C10H18O9
Molecular Weight: 282.25
Purity/Analysis Method: 98.0% (HPLC)
Form: Crystal
Color: White
Specific rotation [a]20/D: -26 deg (C=10, H2O)


Product Categories: Bioscience, Disaccharides
Other Product Names / Synonyms: 4-O-beta-D-Xylopyranosyl-D-xylose

Formula: C₁₀H₁₈O₉
MW: 282.25 g/mol
MDL Number: MFCD00135984
CAS Number: 6860-47-5
https://us.vwr.com/stibo/web/std.lang.all/74/12/28997412.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR