m-Tolylacetonitrile ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 3-Methylbenzyl cyanide(3-Methylphenyl)acetonitrile

X0046-25ML
TCX0046-25MLEA 65 USD
TCX0046-25ML
m-Tolylacetonitrile ≥98.0% (by GC)
m-Tolylacetonitrile

CAS Number: 2947-60-6
MDL Number: MFCD00001914
Molecular Formula: C9H9N
Molecular Weight: 131.18
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 130
Flash Point (°C): 111
Specific Gravity (20/20): 1.00


Other Product Names / Synonyms: 3-Methylbenzyl Cyanide, m-Tolylacetonitrile

Formula: C₉H₉N
MW: 131.18 g/mol
Boiling Pt: 130 °C
Density: 1.00 (20 °C)
Flash Pt: 111 °C
MDL Number: MFCD00001914
CAS Number: 2947-60-6
UN: 3276
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/73/98/28997398.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR