Hydrogen Chloride - Butanol Reagent 5 - 10% for esterification

Danger

X0039-SET
TCX0039-SETEA 212.79 USD
TCX0039-SET
Hydrogen Chloride - Butanol Reagent 5 - 10% for esterification
Hydrogen Chloride - Butanol Reagent

Set of 10 x 1 mL

MDL Number: MFCD00011324
CAS Number: 7647-01-0
UN: 2924
ADR: 3.8,II

Order Now

Specification Test Results

Concentration (Neutralization titration) 5.0 to 10.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR