Boron trifluoride - methanol complex 10 - 20% (by gravimetric analysis) for esterification

Supplier: TCI America

X0036-1SET
TCX0036-SETEA 104.85 USD
TCX0036-SET
Boron trifluoride - methanol complex 10 - 20% (by gravimetric analysis) for esterification
Boron trifluoride methanol complex

Set of 10 x 1 mL

Formula: BF3
MW: 99.85 g/mol
MDL Number: MFCD00071635
CAS Number: 373-57-9
https://us.vwr.com/stibo/web/std.lang.all/73/94/28997394.jpg

Order Now

Specification Test Results

Concentration (Gravimetric Method) 10.0 to 20.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR