m-Phenylenediacetonitrile ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 1,3-Phenylenediacetonitrile

X0002-25G
TCX0002-25GEA 466 USD
TCX0002-25G
m-Phenylenediacetonitrile ≥98.0% (by GC)
m-Phenylenediacetonitrile

CAS Number: 626-22-2
MDL Number: MFCD00001915
Molecular Formula: C10H8N2
Molecular Weight: 156.19
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Color: White
Flash Point (°C): 110
Freezing point (°C): 31


Other Product Names / Synonyms: alpha,alpha'-Dicyano-m-xylene, 1,3-Phenylenediacetonitrile, m-Xylylene Cyanide

Formula: C₁₀H₈N₂
Flash Pt: 110 °C
MDL Number: MFCD00001915
CAS Number: 626-22-2
UN: 3439
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/73/68/28997368.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR