Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Vanillin isobutyrate ≥98.0% (by GC)

Supplier: TCI America
Synonyms: Isobutavan

V0124-25G
TCV0124-25G
Vanillin isobutyrate ≥98.0% (by GC)
CAS Number: 20665-85-4
MDL Number: MFCD00169858
Molecular Formula: C12H14O4
Molecular Weight: 222.24
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 29
Flash Point (°C): 113
Lambda max.: 311 nm (1-Butanol)

Other Product Names / Synonyms: Isobutavan


Formula: C₁₂H₁₄O₄
MW: 222,24 g/mol
Melting Pt: 29 °C
Flash Pt: 113 °C
MDL Number: MFCD00169858
CAS Number: 20665-85-4
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC)
Melting point (°C) 29
Flash Point (°C) 113
Lambda max. 311 nm (1-Butanol)


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty