Vecuronium bromide ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
1-[(2beta,3alpha,5alpha,16beta,17beta)-3,17-Bis(acetyloxy)-2-(1-piperidinyl)androstan-16-yl]-1-methylpiperidinium Bromide
CAS Number: 50700-72-6
MDL Number: MFCD00867762
Molecular Formula: C34H57BrN2O4
Molecular Weight: 637.74
Purity/Analysis Method:
Form: Crystal
Color: White
Melting point (°C): 235
Specific rotation [a]20/D: 32 deg (C=1, 0.05mol/L HCl)
Storage Temperature:
Product Categories: Bioscience, Other Pharmaceutical Ingredients
Other Product Names / Synonyms: 1-[(2beta,3alpha,5alpha,16beta,17beta)-3,17-Bis(acetyloxy)-2-(1-piperidinyl)androstan-16-yl]-1-methylpiperidinium Bromide
Formula:
C₃₄H₅₇BrN₂O₄ MW: 637.74 g/mol Melting Pt: 235 °C |
MDL Number:
MFCD00867762 CAS Number: 50700-72-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...