Bis(cyclopentadienyl)vanadium dichloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Vanadocene dichloride

V0090-1G
TCV0090-1GEA 204.97 USD
TCV0090-1G
Bis(cyclopentadienyl)vanadium dichloride ≥98.0% (by titrimetric analysis)
Bis(cyclopentadienyl)vanadium dichloride

CAS Number: 12083-48-6
Molecular Formula: C10H10Cl2V
Molecular Weight: 252.03
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Color: Green


Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts, Materials Chemistry, Polymerization Catalysts
Other Product Names / Synonyms: Bis(cyclopentadienyl)vanadium Dichloride

Formula: C₁₀H₁₀Cl₂V
MW: 252.04 g/mol
MDL Number: MFCD00014260
CAS Number: 12083-48-6
UN: 3261
ADR: 8,II
https://us.vwr.com/stibo/web/std.lang.all/73/12/28997312.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR