1-(p-Toluenesulfonyl)-1,2,4-triazole ≥98.0% (by HPLC)
Supplier: TCI America
Synonyms:
1-Tosyl-1,2,4-triazole
T3187-5G
T3187-1G
TCT3187-5GEA
102
USD
TCT3187-5G
TCT3187-1G
1-(p-Toluenesulfonyl)-1,2,4-triazole ≥98.0% (by HPLC)
1-(p-Toluenesulfonyl)-1,2,4-triazole
CAS Number: 13578-51-3
MDL Number: MFCD00458591
Molecular Formula: C9H9N3O2S
Molecular Weight: 223.25
Purity/Analysis Method:
Form: Crystal
Color: White
Melting point (°C): 107
Product Categories: Synthetic Organic Chemistry, Condensation & Active Esterification
Other Product Names / Synonyms: 1-Tosyl-1,2,4-triazole
Formula:
C₉H₉N₃O₂S MW: 223.25 g/mol Melting Pt: 107 °C |
MDL Number:
MFCD00458591 CAS Number: 13578-51-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...