Teniposide ≥98.0% (by HPLC)
Supplier: TCI America
Synonyms:
(5R,5aR,8aR,9S)-9-(((2R,4aR,6R,7R,8R,8aS)-7,8-Dihydroxy-2-(thiophen-2-yl)hexahydropyrano[3,2-d][1,3]dioxin-6-yl)oxy)-5-(4-hydroxy-3,5-dimethoxyphenyl)-5,5a,8a,9-tetrahydrofuro[3',4':6,7]naphtho[2,3-d][1,3]dioxol-6(8H)-one
CAS Number: 29767-20-2
MDL Number: MFCD00866516
Molecular Formula: C32H32O13S
Molecular Weight: 656.66
Purity/Analysis Method:
Form: Crystal
Color: White
Melting point (°C): 247
Specific rotation [a]20/D: -111 deg (C=1, CHCl3:MeOH=9:1)
Lambda max.: 283 nm (MeOH)Storage Temperature:
Formula:
C₃₂H₃₂O₁₃S MW: 656.66 g/mol Melting Pt: 247 °C |
MDL Number:
MFCD00866516 CAS Number: 29767-20-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...