N-(2-Thiophenesulfonyl)-L-prolinamide ≥98.0% (by HPLC)

Supplier: TCI America

T3080-100MG
TCT3080-100MGEA 141 USD
TCT3080-100MG
N-(2-Thiophenesulfonyl)-L-prolinamide ≥98.0% (by HPLC)
N-(2-Thiophenesulfonyl)-L-prolinamide

CAS Number: 1089663-51-3
Molecular Formula: C9H12N2O3S2
Molecular Weight: 260.33
Purity/Analysis Method: 98.0% (HPLC)
Form: Crystal
Color: White
Specific rotation [a]20/D: -20.7 deg (C=0.41, MeOH)

Formula: C₉H₁₂N₂O₃S₂
CAS Number: 1089663-51-3
https://us.vwr.com/stibo/web/std.lang.all/66/84/28996684.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR