(R,R)-Ts-DENEB(regR)
Supplier: TCI America
Synonyms:
Chloro[(R,R)-N-[2-[2-(4-methylbenzyloxy)ethyl]amino-1,2-diphenylethyl]-p-toluenesulfonamide]ruthenium(II)
CAS Number: 1333981-84-2
MDL Number: MFCD20922902
Molecular Formula: C31H33ClN2O3RuS
Molecular Weight: 650.20
Form: Crystal
Color: White
Other Product Names / Synonyms: Chloro[(R,R)-N-[2-[2-(4-methylbenzyloxy)ethyl]amino-1,2-diphenylethyl]-p-toluenesulfonamide]ruthenium(II), Chloro[(R,R)-N-[2-(4-methylbenzyloxy)ethyl]-N'-(p-toluenesulfonyl)-1,2-diphenylethylenediamine]ruthenium(II)
Formula:
C₃₁H₃₃ClN₂O₃RuS MW: 650.20 g/mol |
MDL Number:
MFCD20922902 CAS Number: 1333981-84-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...