1,6,7,12-Tetrakis(4-tert-butylphenoxy)-N,N'-bis(2,6-diisopropylphenyl)-3,4,9,10-perylenetetracarboxylic diimide ≥98.0% (by HPLC, total nitrogen)

T3061-200MG
TCT3061-200MGEA 279.72 USD
TCT3061-200MG
1,6,7,12-Tetrakis(4-tert-butylphenoxy)-N,N'-bis(2,6-diisopropylphenyl)-3,4,9,10-perylenetetracarboxylic diimide ≥98.0% (by HPLC, total nitrogen)
1,6,7,12-Tetrakis(4-tert-butylphenoxy)-N,N'-bis(2,6-diisopropylphenyl)-3,4,9,10-perylenetetracarboxylic diimide

CAS Number: 112078-08-7
MDL Number: MFCD15145478
Molecular Formula: C88H90N2O8
Molecular Weight: 1303.69
Purity/Analysis Method: 98.0% (HPLC,N)
Form: Crystal
Color: Yellow Red

Formula: C₈₈H₉₀N₂O₈
MDL Number: MFCD15145478
CAS Number: 112078-08-7
https://us.vwr.com/stibo/web/std.lang.all/66/65/28996665.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR