(1R,4S)-1,2,3,4-Tetrahydro-1,4-epoxynaphthalene-1-carboxylic acid ≥96.0% (by HPLC)

Supplier: TCI America

Synonyms: (R)-THENA

T3001-100MG
TCT3001-100MGEA 386 USD
TCT3001-100MG
(1R,4S)-1,2,3,4-Tetrahydro-1,4-epoxynaphthalene-1-carboxylic acid ≥96.0% (by HPLC)
(1R,4S)-1,2,3,4-Tetrahydro-1,4-epoxynaphthalene-1-carboxylic acid

CAS Number: 1244954-14-0
Molecular Formula: C11H10O3
Molecular Weight: 190.20
Purity/Analysis Method: 96.0% (HPLC)
Form: Crystal
Color: White
Melting point (°C): 139
Specific rotation [a]20/D: -6 deg (C=1, Acetone)


Product Categories: Analytical Science, e.e. / Absolute Configuration Determination (NMR)
Other Product Names / Synonyms: (R)-THENA

Formula: C₁₁H₁₀O₃
MW: 190.20 g/mol
Melting Pt: 139 °C
CAS Number: 1244954-14-0
https://us.vwr.com/stibo/web/std.lang.all/66/19/28996619.jpg

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR