4-(Trifluoromethyl)hydrocinnamic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 3-[4-(Trifluoromethyl)phenyl]propanoic acid

T2927-1G T2927-5G
TCT2927-5GEA 129.92 USD
TCT2927-5G TCT2927-1G
4-(Trifluoromethyl)hydrocinnamic acid ≥98.0% (by titrimetric analysis)
4-(Trifluoromethyl)hydrocinnamic acid

CAS Number: 53473-36-2
MDL Number: MFCD00674032
Molecular Formula: C10H9F3O2
Molecular Weight: 218.18
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 115


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 4-(Trifluoromethyl)hydrocinnamic Acid

Formula: C10H9F3O2
MW: 218.17 g/mol
Melting Pt: 115 °C
MDL Number: MFCD00674032
CAS Number: 53473-36-2
https://us.vwr.com/stibo/web/std.lang.all/65/62/28996562.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR