Thiocolchicoside ≥98.0% (by HPLC)

Supplier: TCI America

T2919-20MG
TCT2919-20MGEA 248 USD
TCT2919-20MG
Thiocolchicoside ≥98.0% (by HPLC)
Thiocolchicoside

CAS Number: 602-41-5
MDL Number: MFCD07995056
Molecular Formula: C27H33NO10S
Molecular Weight: 563.62
Purity/Analysis Method: 98.0% (HPLC)
Form: Crystal
Color: Slightly Pale Yellow
Melting point (°C): 220
Specific rotation [a]20/D: -253 deg (C=1.0, MeOH)
Storage Temperature: 0-10°C

Formula: C₂₇H₃₃NO₁₀S
Melting Pt: 220 °C
MDL Number: MFCD07995056
CAS Number: 602-41-5
https://us.vwr.com/stibo/web/std.lang.all/65/56/28996556.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR