4,4'-Thiodiresorcinol ≥97.0% (by GC)

Synonyms: 4,4'-ThiobisresorcinolBis(2,4-dihydroxyphenyl) sulfide2,2',4,4'-Tetrahydroxydiphenyl sulfide2,2',4,4'-Tetrahydroxydiphenyl sulphideBis(2,4-dihydroxyphenyl) sulphide

T2911-25G
TCT2911-25GEA 309.69 USD
TCT2911-25G
4,4'-Thiodiresorcinol ≥97.0% (by GC)
4,4'-Thiodiresorcinol

CAS Number: 97-29-0
MDL Number: MFCD00045769
Molecular Formula: C12H10O4S
Molecular Weight: 250.27
Purity/Analysis Method: 97.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 180


Product Categories: Synthetic Organic Chemistry, Sulfides
Other Product Names / Synonyms: Bis(2,4-dihydroxyphenyl) Sulfide, 4,4'-Thiobisresorcinol

Formula: C₁₂H₁₀O₄S
Melting Pt: 180 °C
MDL Number: MFCD00045769
CAS Number: 97-29-0
https://us.vwr.com/stibo/web/std.lang.all/65/49/28996549.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR