Triethoxy-1H,1H,2H,2H-heptadecafluorodecylsilane ≥98.0% (by GC)

Synonyms: Triethoxy-1H,1H,2H,2H-perfluorodecylsilane

T2876-5G
TCT2876-5GEA 74.16 USD
TCT2876-5G
Triethoxy-1H,1H,2H,2H-heptadecafluorodecylsilane ≥98.0% (by GC)
Triethoxy-1H,1H,2H,2H-heptadecafluorodecylsilane

CAS Number: 101947-16-4
MDL Number: MFCD00042334
Molecular Formula: C16H19F17O3Si
Molecular Weight: 610.38
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 230
Flash Point (°C): 110
Specific Gravity (20/20): 1.42


Product Categories: Synthetic Organic Chemistry, Fluorous Compounds, Materials Chemistry, Silane Coupling Agents
Other Product Names / Synonyms: Triethoxy-1H,1H,2H,2H-perfluorodecylsilane

Formula: C₁₆H₁₉F₁₇O₃Si
MW: 610.38 g/mol
Boiling Pt: 230 °C
Density: 1.42 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00042334
CAS Number: 101947-16-4
https://us.vwr.com/stibo/web/std.lang.all/65/28/28996528.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR